accessibility menu, dialog, popup

UserName

CAS RN 154-87-0

ClCH3NNSOPOHOOPHOHOOH3CH2NN

CAS RN 154-87-0

IUPAC Name: 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-{[hydroxy(phosphonooxy)phosphoryl]oxy}ethyl)-4-methyl-1,3-thiazol-3-ium chloride
Synonyms: thiamine pyrophosphate cocarboxylase coenzymate 辅羧酶 cocarboxylasum cocarboxilasa thiamine(1+) diphosphate chloride thiamine diphosphate chloride
Molecular Weight (g/mol): 460.76
Molecular Formula: C12H19ClN4O7P2S
InChi Key: YXVCLPJQTZXJLH-UHFFFAOYNA-N
SMILES: [Cl-].CC1=C(CCOP(O)(=O)OP(O)(O)=O)SC=[N+]1CC1=CN=C(C)N=C1N
Quantity 
  • (1)
  • (2)
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results