accessibility menu, dialog, popup

UserName

CAS RN 15364-55-3

IUPAC Name: 1-bromopentacyclo[6.6.6.0²,⁷.0⁹,¹⁴.0¹⁵,²⁰]icosa-2,4,6,9,11,13,15,17,19-nonaene
Synonyms: 1-bromopentacyclo[6.6.6.0²,⁷.0⁹,¹⁴.0¹⁵,²⁰]icosa-2,4,6,9,11,13,15,17,19-nonaene
Molecular Weight (g/mol): 333.23
Molecular Formula: C20H13Br
InChi Key: HSROKCVTEYMWHO-UHFFFAOYSA-N
SMILES: BrC12C3=CC=CC=C3C(C3=CC=CC=C13)C1=CC=CC=C21
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results