CAS RN 153556-42-4
IUPAC Name:
4-bromo-3-fluorobenzoate
Synonyms:
4-bromo-3-fluorobenzoate
Molecular Weight (g/mol):
218
Molecular Formula:
C7H3BrFO2
InChi Key:
RMYOGXPGIDWJLU-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC=C(Br)C(F)=C1
1
–
1
of
1
results
1
Promotions Available
4-Bromo-3-fluorobenzoic acid, 98+%, Thermo Scientific Chemicals
4-Bromo-3-fluorobenzoic acid, 98+%, Thermo Scientific Chemicals
CAS: 153556-42-4 Molecular Formula: C7H4BrFO2 Molecular Weight (g/mol): 219.009 MDL Number: MFCD00672932 InChI Key: RMYOGXPGIDWJLU-UHFFFAOYSA-N Synonym: 3-fluoro-4-bromobenzoic acid,benzoic acid, 4-bromo-3-fluoro,3-fluoro-4-bromo benzoic acid,4-bromo-3-fluoro-benzoic acid,pubchem1316,intermediates-zcf02169,acmc-1c1h6,ksc497g7p,4-bromo-3-fluoro benzoic acid PubChem CID: 2773341 IUPAC Name: 4-bromo-3-fluorobenzoic acid SMILES: C1=CC(=C(C=C1C(=O)O)F)Br