accessibility menu, dialog, popup

UserName

CAS RN 153404-60-5

BrBr

CAS RN 153404-60-5

IUPAC Name: 1-bromo-3-[2-(3-bromophenyl)ethynyl]benzene
Synonyms: 1-bromo-3-[2-(3-bromophenyl)ethynyl]benzene
Molecular Weight (g/mol): 336.03
Molecular Formula: C14H8Br2
InChi Key: WSXUXTVJTIPBAF-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C#CC1=CC(Br)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results