CAS RN 153086-78-3
CAS RN 153086-78-3
IUPAC Name:
tert-butyl N-{2-[2-(2-aminoethoxy)ethoxy]ethyl}carbamate
Synonyms:
tert-butyl N-{2-[2-(2-aminoethoxy)ethoxy]ethyl}carbamate
Molecular Weight (g/mol):
248.32
Molecular Formula:
C11H24N2O4
InChi Key:
OCUICOFGFQENAS-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NCCOCCOCCN
1
–
1
of
1
results
1
N-(tert-Butoxycarbonyl)-2,2'-(ethylenedioxy)diethylamine 97.0+%, TCI America™
CAS: 153086-78-3 Molecular Formula: C11H24N2O4 Molecular Weight (g/mol): 248.323 MDL Number: MFCD03788155 InChI Key: OCUICOFGFQENAS-UHFFFAOYSA-N Synonym: N-Boc-2,2′C-(ethylenedioxy)diethylamine, N-(tert-Butoxycarbonyl)-3,6-dioxa-1,8-octanediamine, N-Boc-3,6-dioxa-1,8-octanediamine, tert-Butyl [2-[2-(2-Aminoethoxy)ethoxy]ethyl]carbamate, [2-[2-(2-Aminoethoxy)ethoxy]ethyl]carbamic Acid tert-Butyl Ester PubChem CID: 9881394 IUPAC Name: tert-butyl N-[2-[2-(2-aminoethoxy)ethoxy]ethyl]carbamate SMILES: CC(C)(C)OC(=O)NCCOCCOCCN