accessibility menu, dialog, popup

UserName

NHO

CAS RN 151666-08-9

IUPAC Name: 3-phenoxypiperidine
Synonyms: 3-phenoxypiperidine
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: QDPNIJKUTWDGMV-UHFFFAOYNA-N
SMILES: C1CNCC(C1)OC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results