accessibility menu, dialog, popup

UserName

CH3OOOHI

CAS RN 15126-06-4

IUPAC Name: methyl 4-hydroxy-3-iodobenzoate
Synonyms: methyl 4-hydroxy-3-iodobenzoate
Molecular Weight (g/mol): 278.05
Molecular Formula: C8H7IO3
InChi Key: PXNOLLHARLSLHY-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(I)=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results