CAS RN 15086-94-9
IUPAC Name:
2',4',5',7'-tetrabromo-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms:
bromeosin solvent red 43 eosin acid eosin Y spirit soluble japan red 223 D and C red no. 21 bromoeosin japan red no. 223 C.I. solvent red 43 eosine acid
Molecular Weight (g/mol):
647.9
Molecular Formula:
C20H8Br4O5
InChi Key:
DBZJJPROPLPMSN-UHFFFAOYSA-N
SMILES:
OC1=C(Br)C=C2C(OC3=C(Br)C(O)=C(Br)C=C3C22OC(=O)C3=CC=CC=C23)=C1Br
1
–
1
of
1
results
1
Tetrabromofluorescein 95.0+%, TCI America™
CAS: 15086-94-9 Molecular Formula: C20H8Br4O5 MDL Number: MFCD00036189 Synonym: Eosin Y