accessibility menu, dialog, popup

UserName

H3COOOOONOO

CAS RN 150321-92-9

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 7-methoxy-2-oxo-2H-chromene-3-carboxylate
Synonyms: 2,5-dioxopyrrolidin-1-yl 7-methoxy-2-oxochromene-3-carboxylate
Molecular Weight (g/mol): 317.25
Molecular Formula: C15H11NO7
InChi Key: JMQAALOXLOSYCQ-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=C(C(=O)ON3C(=O)CCC3=O)C(=O)OC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results