CAS RN 148651-60-9
IUPAC Name:
copper(2+) 6,7,15,16,24,25,33,34-octafluoro-2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19(39),20,22,24,26,28,30,32,34-nonadecaene-37,38-diide
Synonyms:
copper(2+) 6,7,15,16,24,25,33,34-octafluoro-2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19(39),20,22,24,26,28,30,32,34-nonadecaene-37,38-diide
Molecular Weight (g/mol):
720.01
Molecular Formula:
C32H8CuF8N8
InChi Key:
SYOZEEBBXMOLOS-UHFFFAOYSA-N
SMILES:
[Cu++].FC1=CC2=C3[N-]C(N=C4N=C(N=C5N=C(N=C6[N-]C(=N3)C3=CC(F)=C(F)C=C63)C3=CC(F)=C(F)C=C53)C3=CC(F)=C(F)C=C43)=C2C=C1F
1
–
1
of
1
results
1
Copper(II) 2,3,9,10,16,17,23,24-Octafluorophthalocyanine (purified by sublimation) 98.0+%, TCI America™
CAS: 148651-60-9 Molecular Formula: C32H8CuF8N8 Molecular Weight (g/mol): 720.005 InChI Key: SYOZEEBBXMOLOS-UHFFFAOYSA-N Synonym: 2,3,9,10,16,17,23,24-Octafluorophthalocyanine Copper(II), F8CuPc PubChem CID: 53384437 SMILES: C1=C2C(=CC(=C1F)F)C3=NC4=NC(=NC5=NC(=NC6=C7C=C(C(=CC7=C([N-]6)N=C2[N-]3)F)F)C8=CC(=C(C=C85)F)F)C9=CC(=C(C=C94)F)F.[Cu+2]