accessibility menu, dialog, popup

UserName

H3CNH2HBr

CAS RN 14846-47-0

IUPAC Name: octan-1-amine hydrobromide
Synonyms: octylamine hydrobromide
Molecular Weight (g/mol): 210.16
Molecular Formula: C8H20BrN
InChi Key: GIDDQKKGAYONOU-UHFFFAOYSA-N
SMILES: Br.CCCCCCCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results