accessibility menu, dialog, popup

UserName

CAS RN 14783-09-6

NNClCuCl

CAS RN 14783-09-6

IUPAC Name: 1,10-phenanthroline; dichlorocopper
Synonyms: copper chloride; phen
Molecular Weight (g/mol): 314.66
Molecular Formula: C12H8Cl2CuN2
InChi Key: PHECBGBJCLDCMF-UHFFFAOYSA-L
SMILES: Cl[Cu]Cl.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results