accessibility menu, dialog, popup

UserName

FFFNHNFFF

CAS RN 14704-41-7

IUPAC Name: 3,5-bis(trifluoromethyl)-1H-pyrazole
Synonyms: 3,5-bis(trifluoromethyl)-1H-pyrazole
Molecular Weight (g/mol): 204.08
Molecular Formula: C5H2F6N2
InChi Key: NGDDUAYSWPUSLX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=NN1)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)