CAS RN 146137-80-6
IUPAC Name:
2-fluoro-4-methylbenzaldehyde
Synonyms:
2-fluoro-4-methylbenzaldehyde
Molecular Weight (g/mol):
138.14
Molecular Formula:
C8H7FO
InChi Key:
MVDRIMBGRZBWPE-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=O)C(F)=C1
1
–
1
of
1
results
1
Promotions Available
2-Fluoro-4-methylbenzaldehyde, 97%
2-Fluoro-4-methylbenzaldehyde, 97%
CAS: 146137-80-6 Molecular Formula: C8H7FO Molecular Weight (g/mol): 138.141 MDL Number: MFCD03094325 InChI Key: MVDRIMBGRZBWPE-UHFFFAOYSA-N Synonym: 2-fluoro-4-methyl-benzaldehyde,2-fluoro-4-methyl benzaldehyde,benzaldehyde, 2-fluoro-4-methyl,2-fluoro-p-tolualdehyde,pubchem1451,intermediates-zcf02120,acmc-1c24x,ksc494i0d,benzaldehyde, 2-fluoro-4-methyl-9ci PubChem CID: 2778471 IUPAC Name: 2-fluoro-4-methylbenzaldehyde SMILES: CC1=CC(=C(C=C1)C=O)F