accessibility menu, dialog, popup

UserName

CH3NNOCl

CAS RN 14580-22-4

IUPAC Name: 1-(2-chlorophenyl)-3-methyl-4,5-dihydro-1H-pyrazol-5-one
Synonyms: 2-(2-chlorophenyl)-5-methyl-4H-pyrazol-3-one
Molecular Weight (g/mol): 208.65
Molecular Formula: C10H9ClN2O
InChi Key: CWESERWNUIUBJU-UHFFFAOYSA-N
SMILES: CC1=NN(C(=O)C1)C1=CC=CC=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results