accessibility menu, dialog, popup

UserName

CAS RN 145689-34-5

FNF

CAS RN 145689-34-5

IUPAC Name: 2-(2,3-difluorophenyl)acetonitrile
Synonyms: 2-(2,3-difluorophenyl)acetonitrile
Molecular Weight (g/mol): 153.13
Molecular Formula: C8H5F2N
InChi Key: IYRCHGRRMKOSHW-UHFFFAOYSA-N
SMILES: FC1=CC=CC(CC#N)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results