accessibility menu, dialog, popup

UserName

H3CSBr

CAS RN 145543-83-5

IUPAC Name: 2-bromo-3-octylthiophene
Synonyms: 2-bromo-3-octylthiophene
Molecular Weight (g/mol): 275.25
Molecular Formula: C12H19BrS
InChi Key: ISONQKSIWXLJOQ-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=C(Br)SC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results