accessibility menu, dialog, popup

UserName

CAS RN 14489-75-9

CH3HN

CAS RN 14489-75-9

IUPAC Name: methyl[(naphthalen-1-yl)methyl]amine
Synonyms: methyl(naphthalen-1-ylmethyl)amine
Molecular Weight (g/mol): 171.24
Molecular Formula: C12H13N
InChi Key: MQRIUFVBEVFILS-UHFFFAOYSA-N
SMILES: CNCC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results