CAS RN 144851-62-7
IUPAC Name:
2,6-difluoro-3-methoxyaniline
Synonyms:
2,6-difluoro-3-methoxyaniline
Molecular Weight (g/mol):
159.14
Molecular Formula:
C7H7F2NO
InChi Key:
XVLHUQRLEPQGDI-UHFFFAOYSA-N
SMILES:
COC1=C(F)C(N)=C(F)C=C1
1
–
1
of
1
results
1
Promotions Available
2,6-Difluoro-3-methoxyaniline, 97%
2,6-Difluoro-3-methoxyaniline, 97%
CAS: 144851-62-7 Molecular Formula: C7H7F2NO Molecular Weight (g/mol): 159.14 MDL Number: MFCD04115898 InChI Key: XVLHUQRLEPQGDI-UHFFFAOYSA-N Synonym: benzenamine,2,6-difluoro-3-methoxy,acmc-20dww5,2,6-difluoro-m-anisidine,3-amino-2,4-difluoroanisole,2,6-difluoro-3-methoxyphenylamine,benzenamine, 2,6-difluoro-3-methoxy-9ci PubChem CID: 3871278 IUPAC Name: 2,6-difluoro-3-methoxyaniline SMILES: COC1=C(F)C(N)=C(F)C=C1