CAS RN 14464-46-1
CAS RN 14464-46-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
6
of
6
results
1
Clean Loam Soil, MilliporeSigma™ Supelco™
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
2
Clean Clay Loam, MilliporeSigma™ Supelco™
Clean clay loam may be used as a reference material in the recovery of nitro-organic explosives in soil samples using gas chromatography coupled with electron capture detector (GC-ECD).
4
Clean Sand No.4, MilliporeSigma™ Supelco™
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.