accessibility menu, dialog, popup

UserName

CAS RN 144599-95-1

N(S)OHNOHOOCH3CH3CH3OOH3CCH3CH3

CAS RN 144599-95-1

IUPAC Name: (2S)-3-{1-[(tert-butoxy)carbonyl]-1H-indol-3-yl}-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-[1-(tert-butoxycarbonyl)indol-3-yl]propanoic acid|BOC-TRP(BOC)-OH
Molecular Weight (g/mol): 404.46
Molecular Formula: C21H28N2O6
InChi Key: FATGZMFSCKUQGO-HNNXBMFYSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CN(C(=O)OC(C)(C)C)C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results