CAS RN 14441-90-8
IUPAC Name:
5-phenyl-1,2-oxazole-3-carboxylate
Synonyms:
5-phenyl-1,2-oxazole-3-carboxylate
Molecular Weight (g/mol):
188.16
Molecular Formula:
C10H6NO3
InChi Key:
XJYOBHXWBRKOQO-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=NOC(=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
5-Phenylisoxazole-3-carboxylic Acid 98.0+%, TCI America™
CAS: 14441-90-8 Molecular Formula: C10H7NO3 Molecular Weight (g/mol): 189.17 MDL Number: MFCD00464221 InChI Key: XJYOBHXWBRKOQO-UHFFFAOYSA-N Synonym: 5-phenylisoxazole-3-carboxylic acid,5-phenyl-isoxazole-3-carboxylic acid,5-phenyl-3-isoxazolecarboxylic acid,3-isoxazolecarboxylic acid, 5-phenyl,3-isoxazolecarboxylicacid, 5-phenyl,5-phenylisoxazole-3-carboxylicacid,5-phenylisoxazol-3-carboxylic acid PubChem CID: 151916 IUPAC Name: 5-phenyl-1,2-oxazole-3-carboxylic acid SMILES: C1=CC=C(C=C1)C2=CC(=NO2)C(=O)O