accessibility menu, dialog, popup

UserName

CH3OP

CAS RN 1439-36-7

IUPAC Name: 1-(triphenyl-λ⁵-phosphanylidene)propan-2-one
Synonyms: 1-(triphenyl-λ⁵-phosphanylidene)propan-2-one
Molecular Weight (g/mol): 318.36
Molecular Formula: C21H19OP
InChi Key: KAANTNXREIRLCT-UHFFFAOYSA-N
SMILES: CC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)