accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CCH2

CAS RN 143825-84-7

IUPAC Name: 4,4,5,5-tetramethyl-2-(1-phenylethenyl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(1-phenylethenyl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 230.11
Molecular Formula: C14H19BO2
InChi Key: RMGBWPMWUZSIMH-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C(=C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results