accessibility menu, dialog, popup

UserName

PtNNNNKKH2OH2OH2O

CAS RN 14323-36-5

IUPAC Name: dipotassium tetracyanoplatinumbis(ylium) trihydrate
Synonyms: dipotassium tetracyanoplatinumbis(ylium) trihydrate
Molecular Weight (g/mol): 431.4
Molecular Formula: C4H6K2N4O3Pt
InChi Key: KBALGZKVNFJSRU-UHFFFAOYSA-N
SMILES: O.O.O.[K+].[K+].N#C[Pt++](C#N)(C#N)C#N
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results