accessibility menu, dialog, popup

UserName

CAS RN 14312-89-1

IUPAC Name: methyl 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptanoate
Synonyms: methyl 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptanoate
Molecular Weight (g/mol): 378.09
Molecular Formula: C8H3F13O2
InChi Key: JHROQORAJUWVCD-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results