CAS RN 1421-65-4
IUPAC Name:
hydrogen methyl 2-amino-3-(3,4-dihydroxyphenyl)propanoate chloride
Synonyms:
hydrogen methyl 2-amino-3-(3,4-dihydroxyphenyl)propanoate chloride
Molecular Weight (g/mol):
247.68
Molecular Formula:
C10H14ClNO4
InChi Key:
WFGNJLMSYIJWII-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].COC(=O)C(N)CC1=CC=C(O)C(O)=C1
1
–
1
of
1
results
1
L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride, 95%
CAS: 1421-65-4 Molecular Formula: C10H14ClNO4 Molecular Weight (g/mol): 247.68 InChI Key: WFGNJLMSYIJWII-UHFFFAOYNA-N Synonym: s-methyl 2-amino-3-3,4-dihydroxyphenyl propanoate hydrochloride,l-3,4-dihydroxyphenylalanine methyl ester hydrochloride,methyl l-dopa hydrochloride,melevodopa hydrochloride,unii-jfu1a8866v,l-dopa methyl ester hydrochloride,levodopa methyl ester hydrochloride,l-tyrosine, 3-hydroxy-, methyl ester, hydrochloride,dsstox_cid_25763,dsstox_rid_81105 PubChem CID: 10131132 SMILES: [H+].[Cl-].COC(=O)C(N)CC1=CC=C(O)C(O)=C1