accessibility menu, dialog, popup

UserName

H3CNHCH3

CAS RN 142-84-7

IUPAC Name: dipropylamine
Synonyms: dipropylamine
Molecular Weight (g/mol): 101.19
Molecular Formula: C6H15N
InChi Key: WEHWNAOGRSTTBQ-UHFFFAOYSA-N
SMILES: CCCNCCC
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)