CAS RN 142-18-7
IUPAC Name:
2,3-dihydroxypropyl dodecanoate
Synonyms:
1-monomyristin C12-18 acid triglyceride monolaurin
Molecular Weight (g/mol):
274.4
Molecular Formula:
C15H30O4
InChi Key:
ARIWANIATODDMH-UHFFFAOYNA-N
SMILES:
CCCCCCCCCCCC(=O)OCC(O)CO
1
–
1
of
1
results
1
Monolaurin 98.0+%, TCI America™
CAS: 142-18-7 Molecular Formula: C15H30O4 Molecular Weight (g/mol): 274.401 MDL Number: MFCD00037815 InChI Key: ARIWANIATODDMH-UHFFFAOYSA-N Synonym: monolaurin,1-monolaurin,lauricidin,glyceryl laurate,glyceryl monolaurate,1-glyceryl laurate,glycerol 1-laurate,1-monolauroyl-rac-glycerol,1-monododecanoylglycerol,glycerol monolaurate PubChem CID: 14871 ChEBI: CHEBI:75539 IUPAC Name: 2,3-dihydroxypropyl dodecanoate SMILES: CCCCCCCCCCCC(=O)OCC(CO)O