accessibility menu, dialog, popup

UserName

CH3NCH3CH3H3C

CAS RN 14190-16-0

IUPAC Name: tetramethylazanium
Synonyms: tetramethylammonium ion whelk poison tetraamethylammonium acetate whelk poison (substance)
Molecular Weight (g/mol): 74.15
Molecular Formula: C4H12N
InChi Key: QEMXHQIAXOOASZ-UHFFFAOYSA-N
SMILES: C[N+](C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results