accessibility menu, dialog, popup

UserName

CAS RN 1414-45-5

IUPAC Name: 6-amino-2-{2-[2-(2-{2-[2-({7-[6-amino-2-(2-{2-[(21-{6-amino-2-[(10-{15-[(2Z)-2-(2-amino-3-methylpentanamido)but-2-enamido]-12-(butan-2-yl)-9-methylidene-6-(2-methylpropyl)-5,8,11,14-tetraoxo-1-thia-4,7,10,13-tetraazacyclohexadecane-3-amido}-9-methyl-1,4,11-trioxo-tetradecahydropyrrolo[2,1-i]1-thia-4,7,10-triazacyclotridecan-6-yl)formamido]hexanamido}-15,22-dimethyl-12-(2-methylpropyl)-9-[2-(methylsulfanyl)ethyl]-5,8,11,14,17,20-hexaoxo-1-thia-4,7,10,13,16,19-hexaazacyclodocosan-3-yl)formamido]-3-carbamoylpropanamido}-4-(methylsulfanyl)butanamido)hexanamido]-14-[(1H-imidazol-5-yl)methyl]-4,8,20-trimethyl-3,6,12,15,21-pentaoxo-9,19-dithia-2,5,13,16,22-pentaazabicyclo[9.9.2]docosan-17-yl}formamido)-3-hydroxypropanamido]-3-methylpentanamido}-3-(1H-imidazol-5-yl)propanamido)-3-methylbutanamido]prop-2-enamido}hexanoic acid
Synonyms: nisin
Molecular Weight (g/mol): 3354.09
Molecular Formula: C143H230N42O37S7
InChi Key: NVNLLIYOARQCIX-GSJOZIGCNA-N
SMILES: CCC(C)C(N)C(=O)N\C(=C/C)C(=O)NC1CSCC(NC(=O)C(CC(C)C)NC(=O)C(=C)NC(=O)C(NC1=O)C(C)CC)C(=O)NC1C(C)SCC(NC(=O)CNC(=O)C2CCCN2C1=O)C(=O)NC(CCCCN)C(=O)NC1C(C)SCC(NC(=O)CNC(=O)C(CCSC)NC(=O)C(CC(C)C)NC(=O)C(C)NC(=O)CNC1=O)C(=O)NC(CC(N)=O)C(=O)NC(CCSC)C(=O)NC(CCCCN)C(=O)NC1C(C)SCC2NC(=O)C(NC(=O)C(C)NC1=O)C(C)SCC(NC(=O)C(CC1=CN=CN1)NC2=O)C(=O)NC(CO)C(=O)NC(C(C)CC)C(=O)NC(CC1=CN=CN1)C(=O)NC(C(C)C)C(=O)NC(=C)C(=O)NC(CCCCN)C(O)=O
Color 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Type 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results