accessibility menu, dialog, popup

UserName

CAS RN 141029-75-6

IUPAC Name: 7-octyl-3,11-dithia-7-azatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Synonyms: 7-octyl-3,11-dithia-7-azatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Molecular Weight (g/mol): 291.47
Molecular Formula: C16H21NS2
InChi Key: GGCJLBJHTUIOES-UHFFFAOYSA-N
SMILES: CCCCCCCCN1C2=C(SC=C2)C2=C1C=CS2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results