accessibility menu, dialog, popup

UserName

NH2H2NN

CAS RN 141-86-6

IUPAC Name: pyridine-2,6-diamine
Synonyms: 2,6-diaminopyridine
Molecular Weight (g/mol): 109.13
Molecular Formula: C5H7N3
InChi Key: VHNQIURBCCNWDN-UHFFFAOYSA-N
SMILES: NC1=CC=CC(N)=N1
Boiling Point 
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)