CAS RN 140164-85-8
IUPAC Name:
2-[methyl(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]acetyl chloride
Synonyms:
2-[methyl(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]acetyl chloride
Molecular Weight (g/mol):
270.63
Molecular Formula:
C9H7ClN4O4
InChi Key:
BLKGIXBLRWZLKS-UHFFFAOYSA-N
SMILES:
CN(CC(Cl)=O)C1=CC=C(C2=NON=C12)[N+]([O-])=O
1
–
1
of
1
results
1
NBD-COCl 92.0+%, TCI America™
CAS: 140164-85-8 Molecular Formula: C9H7ClN4O4 Molecular Weight (g/mol): 270.629 MDL Number: MFCD02093432 InChI Key: BLKGIXBLRWZLKS-UHFFFAOYSA-N Synonym: 4-(N-Chloroformylmethyl-N-methylamino)-7-nitro-2,1,3-benzoxadiazole PubChem CID: 23146122 IUPAC Name: 2-[methyl-(4-nitro-2,1,3-benzoxadiazol-7-yl)amino]acetyl chloride SMILES: CN(CC(=O)Cl)C1=CC=C(C2=NON=C12)[N+](=O)[O-]