accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OFFZn

CAS RN 13986-18-0

IUPAC Name: zinc(2+) tetrahydrate difluoride
Synonyms: zinc(2+) tetrahydrate difluoride
Molecular Weight (g/mol): 175.44
Molecular Formula: F2H8O4Zn
InChi Key: OEUUYPRXVRBIFO-UHFFFAOYSA-L
SMILES: O.O.O.O.[F-].[F-].[Zn++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results