CAS RN 139264-17-8
CAS RN 139264-17-8
IUPAC Name:
(4R)-4-({3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}methyl)-1,3-oxazolidin-2-one
Synonyms:
zomigon zomig rapimelt AscoTop ZMT
Molecular Weight (g/mol):
287.36
Molecular Formula:
C16H21N3O2
InChi Key:
ULSDMUVEXKOYBU-CYBMUJFWSA-N
SMILES:
CN(C)CCC1=CNC2=CC=C(C[C@@H]3COC(=O)N3)C=C12
1
–
1
of
1
results
1
Zolmitriptan 98.0+%, TCI America™
CAS: 139264-17-8 Molecular Formula: C16H21N3O2 Molecular Weight (g/mol): 287.36 MDL Number: MFCD00871503 InChI Key: ULSDMUVEXKOYBU-CYBMUJFWSA-N Synonym: zolmitriptan,zomig,zomigoro,zomigon,flezol,zomig-zmt,zomig nasal spray,zomig zmt,zolmitriptane,zolmitriptanum PubChem CID: 60857 ChEBI: CHEBI:10124 IUPAC Name: (4R)-4-({3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}methyl)-1,3-oxazolidin-2-one SMILES: CN(C)CCC1=CNC2=CC=C(C[C@@H]3COC(=O)N3)C=C12