accessibility menu, dialog, popup

UserName

HONHOSHNCl

CAS RN 138977-28-3

IUPAC Name: N-[2-(4-chlorophenyl)ethyl]-7,8-dihydroxy-2,3,4,5-tetrahydro-1H-2-benzazepine-2-carbothioamide
Synonyms: capsazepine
Molecular Weight (g/mol): 376.9
Molecular Formula: C19H21ClN2O2S
InChi Key: DRCMAZOSEIMCHM-UHFFFAOYSA-N
SMILES: OC1=CC2=C(CN(CCC2)C(=S)NCCC2=CC=C(Cl)C=C2)C=C1O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results