accessibility menu, dialog, popup

UserName

CAS RN 138906-41-9

IUPAC Name: [3,4-bis(acetyloxy)-5-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-6-(4-methoxyphenoxy)oxan-2-yl]methyl acetate
Synonyms: [3,4-bis(acetyloxy)-5-(1,3-dioxoisoindol-2-yl)-6-(4-methoxyphenoxy)oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 541.51
Molecular Formula: C27H27NO11
InChi Key: YKSPBIYKEHIGHE-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C2N2C(=O)C3=CC=CC=C3C2=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results