accessibility menu, dialog, popup

UserName

ONOOBr

CAS RN 13831-59-9

IUPAC Name: 1-(2-bromoethoxy)-3-nitrobenzene
Synonyms: 1-(2-bromoethoxy)-3-nitrobenzene
Molecular Weight (g/mol): 246.06
Molecular Formula: C8H8BrNO3
InChi Key: QBUSKXLDUNPEMZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(OCCBr)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)