accessibility menu, dialog, popup

UserName

CAS RN 13807-84-6

IUPAC Name: (2-phenoxyphenyl)methanol
Synonyms: (2-phenoxyphenyl)methanol
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: VMZBMTWFHYYOIN-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1OC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)