accessibility menu, dialog, popup

UserName

CAS RN 137255-40-4

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-{[3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]carbamoyl}propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-{[3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]carbamoyl}propanoic acid
Molecular Weight (g/mol): 435.43
Molecular Formula: C17H29N3O10
InChi Key: YVKMLCHQJOPFTE-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(O)C(O)C(CO)OC1NC(=O)CC(NC(=O)OC(C)(C)C)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results