accessibility menu, dialog, popup

UserName

CAS RN 13725-33-2

IUPAC Name: dipotassium 3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate
Synonyms: dipotassium 3,3'-dioxo-1H,1'H-[2,2'-biindolylidene]-5,5'-disulfonate
Molecular Weight (g/mol): 498.56
Molecular Formula: C16H8K2N2O8S2
InChi Key: PLXHLMUZUKUFAR-UHFFFAOYSA-L
SMILES: [K+].[K+].[O-]S(=O)(=O)C1=CC2=C(NC(C2=O)=C2NC3=C(C=C(C=C3)S([O-])(=O)=O)C2=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results