accessibility menu, dialog, popup

UserName

VVOSOOOOSOOOOSOOO

CAS RN 13701-70-7

IUPAC Name: divanadium trisulfate
Synonyms: vanadium(III) sulfate
Molecular Weight (g/mol): 390.05
Molecular Formula: O12S3V2
InChi Key: OYCGXLKTCYDJNJ-UHFFFAOYSA-H
SMILES: [V].[V].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Grade 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results