CAS RN 13686-66-3
IUPAC Name:
(diethylamino)dimethylsilyl
Synonyms:
(diethylamino)dimethylsilyl
Molecular Weight (g/mol):
130.29
Molecular Formula:
C6H16NSi
InChi Key:
ADTGAVILDBXARD-UHFFFAOYSA-N
SMILES:
CCN(CC)[Si](C)C
1
–
1
of
1
results
1
Promotions Available
Dimethylsilyldiethylamine, 95%
Dimethylsilyldiethylamine, 95%
CAS: 13686-66-3 Molecular Formula: C6H16NSi Molecular Weight (g/mol): 130.29 MDL Number: MFCD00048523 InChI Key: ADTGAVILDBXARD-UHFFFAOYSA-N Synonym: dimethylsilyldiethylamine,n,n-diethyl-1,1-dimethylsilylamine,diethylamino dimethylsilyl,silanamine, n,n-diethyl-1,1-dimethyl,silanamine,n,n-diethyl-1,1-dimethyl,diethylamino dimethyl silicon,diethylaminodimethylsilane,n,n-diethyl-1,1-dimethylsilanamine PubChem CID: 6335300 IUPAC Name: (diethylamino)dimethylsilyl SMILES: CCN(CC)[Si](C)C