CAS RN 13678-68-7
IUPAC Name:
1-{[(furan-2-yl)methyl]sulfanyl}ethan-1-one
Synonyms:
1-[(furan-2-ylmethyl)sulfanyl]ethanone
Molecular Weight (g/mol):
156.2
Molecular Formula:
C7H8O2S
InChi Key:
LQOUTUIIYXYBQW-UHFFFAOYSA-N
SMILES:
CC(=O)SCC1=CC=CO1
1
–
1
of
1
results
1
S-Furfuryl Thioacetate 97.0+%, TCI America™
CAS: 13678-68-7 Molecular Formula: C7H8O2S Molecular Weight (g/mol): 156.199 MDL Number: MFCD00010083 InChI Key: LQOUTUIIYXYBQW-UHFFFAOYSA-N Synonym: s-furfuryl thioacetate,furfuryl thioacetate,furfurylthiol acetate,ethanethioic acid, s-2-furanylmethyl ester,s-furfuryl ethanethioate,acetic acid, thio-, s-furfuryl ester,s-2-furanylmethyl ethanethioate,fema no. 3162,2-furfurylthiol acetate,thioacetic acid s-furfuryl ester PubChem CID: 61660 IUPAC Name: S-(furan-2-ylmethyl) ethanethioate SMILES: CC(=O)SCC1=CC=CO1