accessibility menu, dialog, popup

UserName

CH3OHH3C

CAS RN 13651-14-4

IUPAC Name: (2,3-dimethylphenyl)methanol
Synonyms: (2,3-dimethylphenyl)methanol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: ZQQIVMXQYUZKIQ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(CO)=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results