accessibility menu, dialog, popup

UserName

CH3OONNH3COOH3CClCl

CAS RN 135590-91-9

IUPAC Name: 3,5-diethyl 1-(2,4-dichlorophenyl)-5-methyl-4,5-dihydro-1H-pyrazole-3,5-dicarboxylate
Synonyms: mefenpyr-diethyl
Molecular Weight (g/mol): 373.23
Molecular Formula: C16H18Cl2N2O4
InChi Key: OPGCOAPTHCZZIW-UHFFFAOYNA-N
SMILES: CCOC(=O)C1=NN(C2=C(Cl)C=C(Cl)C=C2)C(C)(C1)C(=O)OCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results