accessibility menu, dialog, popup

UserName

CH3SOOOH3CNNSOCH3

CAS RN 1352947-66-0

IUPAC Name: 1-(6-methanesulfinylhexyl)-3-methyl-1H-imidazol-3-ium 4-methylbenzene-1-sulfonate
Synonyms: 3-(6-methanesulfinylhexyl)-1-methylimidazol-1-ium tosylate
Molecular Weight (g/mol): 400.55
Molecular Formula: C18H28N2O4S2
InChi Key: YITFXJWLFMQQRC-UHFFFAOYNA-M
SMILES: CC1=CC=C(C=C1)S([O-])(=O)=O.C[N+]1=CN(CCCCCCS(C)=O)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results