CAS RN 135-55-7
IUPAC Name:
sodium 7-hydroxynaphthalene-2-sulfonate
Synonyms:
sodium 7-hydroxynaphthalene-2-sulfonate
Molecular Weight (g/mol):
246.21
Molecular Formula:
C10H7NaO4S
InChi Key:
IVXSQYUPCUOMRV-UHFFFAOYSA-M
SMILES:
[Na+].OC1=CC=C2C=CC(=CC2=C1)S([O-])(=O)=O
1
–
1
of
1
results
1
Sodium 2-Naphthol-7-sulfonate Hydrate 98.0+%, TCI America™
CAS: 135-55-7 Molecular Formula: C10H7NaO4S Molecular Weight (g/mol): 246.212 MDL Number: MFCD00021625 InChI Key: IVXSQYUPCUOMRV-UHFFFAOYSA-M Synonym: Cassella′Cs Acid Sodium Salt, F Acid Sodium Salt, 7-Hydroxy-2-naphthalenesulfonic Acid Sodium Salt, Sodium 7-Hydroxy-2-naphthalenesulfonate, 2-Naphthol-7-sulfonic Acid Sodium Salt PubChem CID: 23667647 IUPAC Name: sodium;7-hydroxynaphthalene-2-sulfonate SMILES: C1=CC(=CC2=C1C=CC(=C2)S(=O)(=O)[O-])O.[Na+]