accessibility menu, dialog, popup

UserName

CAS RN 13478-18-7

CAS RN 13478-18-7

IUPAC Name: molybdenum(3+) trichloride
Synonyms: molybdenum(III) chloride
Molecular Weight (g/mol): 202.3
Molecular Formula: Cl3Mo
InChi Key: ZSSVQAGPXAAOPV-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Mo+3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results